missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Agarose, high gelling temperature, Molecular Biology Reagent
181.45 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
High gel strength allows gel formation at low concentrations. Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- High gel strength allows gel formation at low concentrations
- Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder |
Color | White |
Quantity | 100 g |
Sensitivity | Hygroscopic; Moisture sensitive |
Chemical Name or Material | Agarose, high gelling temperature, Molecular Biology Reagent |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'