missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose low EEO, for electrophoresis, (Mr <= 0.12)

Product Code. 10072040 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
10g
25g
This item is not returnable. View return policy

Product Code. 10072040

Brand: Thermo Scientific Chemicals 159820100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9012-36-6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
MDL Number MFCD00081294

Specifications

Physical Form Fine Crystalline Powder
Color White
Quantity 10 g
Electroendosmosis (EEO) electroendosmosis (-Mr): 0.07 to 0.12
Gelation Temperature 40 to 43°C (1.5% sol.)
Gel Strength >1800g/cm2 (1.5% gel)
Infrared Spectrum Authentic
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in hot water. Other solubilities: insoluble in most common organic solvents
Grade Electrophoresis
Chemical Name or Material Agarose low EEO
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.