missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose (Medium-EEO/Protein Electrophoresis Grade), Fisher BioReagents
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
Description
Ideal alternative to polyacrylamide for serum protein electrophoresis. Immunoelectrophoresis. Electrophoresis of nucleic acids.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder/Solid |
CAS | 9012-36-6 |
Color | Cream |
Quantity | 100 g |
DNase | DNase free |
Electroendosmosis (EEO) | 0.16 to 0.19 |
Molecular Formula | C12H18O9 |
Gelation Temperature | 34.5° to 37.5°C |
Gel Strength | 1000g/cm2min. |
Formula Weight | 306.12 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'