Learn More
MP Biomedicals Agarose, Molecular Biology Grade
Essential component in chemical separation techniques.
Brand: MP Biomedicals 11AGAH0100
140.10 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : CAS Number : 9012-36-6 Weight : 0.10000kg
Description
- Standard agarose is a purified linear galactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
- Key applications: Electrophoresis, separation of DNA/RNA fragments from 400 to 8000 bp.
- Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.
Specifications
Typical working solution: 1.5% gel | |
Agarose gel powder | |
88°C ±1.5°C (1.5% gel) | |
Room temperature |
White | |
Molecular biology, electrophoresis, separation of DNA/RNA fragments from 400 to 8000 bp | |
1 x 100 g |