missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose (Protein Electrophoresis Grade), Fisher BioReagents™
638.35 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Description
Agarose (Protein Electrophoresis Grade)
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Powder/Solid |
Color | Cream |
Quantity | 500 g |
DNase | DNase free |
Electroendosmosis (EEO) | 0.1 max. |
Gelation Temperature | 34° to 45°C |
Gel Strength | 800g/cm2min. |
Formula Weight | 306.12 |
Grade | DNase- and RNase-Free |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'