missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Agarose Standard

Agarose Standard, Molecular Biology Grade, White Powder

Brand:  MP Biomedicals™ 11AGAH0250

326.58 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 11496950

  • £383.00 / 250g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Standard agarose for analytical electrophoresis
  • Low EEO for complete resolution of bands >500 bp
  • Highly purifi ed, strict quality test procedures
  • Tested for transfer to membrane
  • QA-AgaroseTM Standard is a standard gelling temperature agarose with very low EEO (electroendosmosis) values giving complete resolution
  • of bands greater than 500 bp.
  • This agarose is recommended for analysis when sharp resolution of bands >500 bp is required.
  • The product is quality tested on a lot basis to ensure the absence of DNases and RNases and for the absence of inhibitors to restriction enzymes.
  • Each batch of agarose must pass a DNA transfer test whereby full DNA transfer must occur from gel to membrane in a 1 hour vacuum transfer procedure.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

9012-36-6
White
Powder
89°C (1.5%gel)
15 - 30°C
Standard agarose is a purified lineargalactan hydrocolloid isolated from agar or agar-bearing marine algae with an Electroendoosmosis (EEO) value of 0.12 making it an essential component in chemical separation techniques.
Agarose
Electrophoresis; Separation of DNA/RNA fragments from 400 to 8000 bp
250 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Agarose Standard > 250g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.