missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agarose Standard, MP Biomedicals™
560.90 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
500 g
Description
This Standard Agarose may be used at concentrations between 0.8% and 2%. Southern and Northern transfers can be performed efficiently from them.
Specifications
CAS | 9012-36-6 |
Melting Point | ∼89°C (1.5% gel) |
Quantity | 500 g |
Gel Strength | ≥1200g/cm3 |
Synonym | Galactan Hydrocolloid, 3,6-Anhydro-α-L-galacto-β-D-galactan, FastLane agarose, Indubiose A4, NuSieve GTG, Odigose, Seakem, Sepharose |
Solubility Information | Soluble in water (1.5 g plus 100ml) |
Grade | Molecular Biology |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'