missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Agarose Ultra Pure

Ideal for everyday analysis of nucleic acids by gele electrophoresis or blotting (Northern or Southern) and is also suitable for protein applications.

£213.00 - £863.00

Specifications

For Use With (Application) PCR,Electrophoresis
Storage Requirements Store at room temperature Light Sensitive
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16042565
View Documents
Abnova™
D0021.100g
100 g
£213.00
100g
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
16052565
View Documents
Abnova™
D0021.500g
500 g
£863.00
500g
Estimated Shipment: 18-12-2024
to see stock.
Add to basket
 
Description

Description

No background fluorescence has been detected by ethidium bromide assay. DNase/RNase-free.
Specifications

Specifications

PCR,Electrophoresis
Store at room temperature Light Sensitive
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 100GR Agarose Ultra Pure

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P233-Keep container tightly closed.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P404-Store in a closed container.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium phosphate dibasic, potassium phosphate monobasic

SDS
Documents

Documents

Product Certifications