missing translation for 'onlineSavingsMsg'
Learn More

SPEX™ Agate Grinding Ball

SPEX™ Agate Grinding Ball is a replacement ball for use with 8014 and 3118 Agate Grinding Vial Set.

Brand:  SPEX™ 3118A

 View more versions of this product

Product Code. 15310532

Quantity Price
1 £24.75 / Each
10 £20.57 / Each
Estimated Shipment: 25-10-2024
to see stock.
Explore more special offers
Add to basket

This item is not returnable. View return policy

Description

Description

  • Ideal for grinding and mixing samples when organic and metallic contamination are equally undesirable
  • Extremely high resistance to abrasion and very long wearing withmoderate efficiency
  • Handmade from flawless Brazilian agate
  • Typical composition: Major element – Si; Minor elements – Al, Na, Fe, K, Ca, Mg
  • Hardness: Mohs 6-7; Knoop 550-800
Specifications

Specifications

Grinding Balls
6.35 mm
3118 Agate Grinding Vial Set
Agate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers