Learn More
RETSCH Agate Grinding Balls
Brand: RETSCH 05.368.0111
Code : 50
Additional Details : Weight : 0.01500kg
Specifications
Grinding Balls | |
15 mm | |
PM 100, PM 200, PM 400 Planetary Ball Mills | |
Polished Agate |