missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beckman Coulter™ Agencourt AMPure XP
Click to view available options
Volume:
5mL
60mL
450mL
Unit Size:
450mL
Each
Description
- SPRI paramagnetic bead-based chemistry
- Removes contaminants (dNTPs, salts, primers, primer dimers)
- High recovery of amplicons (>100 bp) ensures that critical data is not lost
- Trusted and recommended reagent for a variety of NGS workflows
- Compatible with manual and automated processing
- Predictable and consistent size selection
- Manual cleanups using AMPure XP require less than 30 minutes
- AMPure XP is automated on all Biomek platforms for higher throughput
Specifications
Specifications
| Composition | AMPure XP, Speed Beads (Mag Carboxy) |
| Content And Storage | 2-8°C |
| Form | Liquid |
| For Use With (Equipment) | Automated 96- or 384-well formats, e.g. Biomek platforms |
| pH | 8.0 to 8.4 |
| For Use With (Application) | Standard PCR, Real Time PCR (qPCR), Next-Generation Sequencing (NGS) |
| Sample Type | DNA |
| Volume | 5mL |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction