Learn More
Brady™ Aggressive Adhesive Multipurpose Nylon Labels
The B-499 nylon cloth label is made of tough and versatile, general-purpose label material. Print almost anywhere with a Brady™ portable label printer.
Brand:  Brady™ 170934
49.71 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 17871809
  • £55.75 / Pack of 160
Explore more special offers
Description
Description
  • Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike
Specifications
Specifications
UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863)
Permanent Acrylic
Matte
Nylon Cloth
Non-sterile
BMP51, M511
-40°F to +194°F
1.25 x 1.5 in.
1.5 in.
M5-132-499
Black on White
Die-cut
Labels
Component and Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking, Wire and Cable Labeling
160 Labels/Cartridge
0.0065 in.
1.25 in.
3.8 cm