Learn More
Brady™ Aggressive Adhesive Multipurpose Nylon Vial and Tube Labels
The B-499 nylon cloth label is made of tough and versatile, general-purpose label material. Print almost anywhere with a Brady™ portable label printer.
Brand:  Brady™ 170799
 View more versions of this product
Product Code. 17980740
  • £43.45 / Pack of 240
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for labeling tightly-curved surfaces such as wires and cables, as well as general-purpose lab, equipment panels, faceplates and terminal blocks
  • Designed to withstand extreme temperatures, chemicals, greases, oils, cleaners and detergents
  • Permanent adhesive labels that stick to smooth, rough and powder-coated surfaces alike
Specifications
Specifications
UL Recognized to UL969, CSA to C22.2 No. 0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863)
White
Die-cut
Labels
Component and Equipment Labeling, Electrical Labeling, General Identification, Laboratory Labeling, Warehouse Marking, Wire and Cable Labeling
240 Labels/Cartridge
0.0065 in.
0.5 in.
1.3 cm
Permanent Acrylic
Matte
Nylon Cloth
Non-sterile
BMP41, BMP51, M511
-40°F to +194°F
0.5 x 0.5 in.
0.5 in.
M4-83-499
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Brady™ Aggressive Adhesive Multipurpose Nylon Vial and Tube Labels > Dimensions (LxW): 0.5 x 0.5 in.; For Use With (Equipment): BMP41, BMP51, M511; Format: Die-cut