missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ AGI-5198, 95%
Brand: Thermo Scientific™ 467921000
306.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
AGI-5198 | |
N-[2-(Cyclohexylamino)-1-(2-methylphenyl)-2-oxoethyl]-N-(3-fluorophenyl)-2-methyl-1 H-imidazole-1-acetamide | |
C27 H31 F N4 O2 | |
Solid | |
95% |
1355326-35-0 | |
100 mg | |
462.56 | |
Reagent |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
344356 | Certificate of Analysis | 20/07/2020 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ AGI-5198, 95% > 100 mg
Spot an opportunity for improvement?Share a Content Correction