missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ AgilePulse™ In Vivo ID (Intra-Dermal) System
AgilePulse™ In Vivo ID (Intra-Dermal) System provides maximum efficiency DNA and RNA delivery to skin, muscle, and tumor tissues.
Brand: BTX™ 470400N
This item is not returnable.
View return policy
Description
- Used for animal immunization, vaccine development and gene therapy, provides an intra-muscular, intra-dermal or intra-tumor electroporation solution to produce maximum transfection efficiency
- Simple user interface
- Pulse Agile Advantage
- Uniform, reliable electroporation
- Simple, Fast Treatment
Specifications
Vaccine Development, Intra-Dermal Gene Delivery, Intra-Tumor Gene Delivery | |
BTX Electroporation | |
50 to 1000 V | |
Touchscreen | |
40 cm | |
20 cm | |
50°F to 104°F | |
10°C to 40°C | |
25 lb. | |
12.6 in. | |
Non-sterile | |
Touch Screen Display, Footswitch |
5 kHz to 1 Hz | |
Electroporation System | |
AgilePulse In Vivo ID (Intra-Dermal) System | |
15.7 in. | |
7.9 in. | |
AgilePulse™ ID Generator, Handle for Parallel Needle Array (47-0000), 4-needle Parallel Needle Array (47-0040 - 3 Pk.), 6-needle Parallel Needle Array (47-0050 - 3 Pk.) and ID Software | |
0.200 to 1000 ms | |
0.050 to 10 ms | |
11.3 kg | |
32 cm | |
Electrode Connector Cable | |
Square Wave |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BTX™ AgilePulse™ In Vivo ID (Intra-Dermal) System > AgilePulse In Vivo ID (Intra-Dermal) System
Intended for Research use only. Not for use in diagnostic or clinical procedures.
Spot an opportunity for improvement?Share a Content Correction