missing translation for 'onlineSavingsMsg'
Learn More

AGN 192403 hydrochloride, Tocris Bioscience™

Product Code. 18025472
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

I1 selective ligand

I1 imidazoline binding site selective ligand with a potency at I1 comparable to moxonidine, but devoid of affinity for adrenoceptors and the I2 binding site. Interestingly, in animal models, this compound causes none of the physiological responses associated with the I1 binding site.

Chemical Identifiers

CAS 1021868-90-5
Molecular Formula C10H20ClN
Molecular Weight (g/mol) 189.727
InChI Key KSTGYMXUFHCTSM-UHFFFAOYSA-N
Synonym agn 192403 hydrochloride, 3-isopropylbicyclo 2.2.1 heptan-2-amine hydrochloride, +/--2-endo-amino-3-exo-isopropylbicyclo 2.2.1 heptane hydrochloride, 2-endo,3exo-3-methylethyl-bicyclo 2.2.1 heptan-2-amine hydrochloride
PubChem CID 11957452
IUPAC Name 2-propan-2-ylbicyclo[2.2.1]heptan-3-amine;hydrochloride
SMILES CC(C)C1C2CCC(C2)C1N.Cl

Specifications

Quantity 10 mg
Formula Weight 189.73
Percent Purity >98%
Chemical Name or Material AGN 192403 hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'