Learn More
Applied Biosystems™ AgPath-ID™ One-Step RT-PCR Reagents with manual
AgPath-ID™ One-Step RT-PCR Reagents are designed for sensitive, robust amplification of RNA targets using a rapid single-tube TaqMan™ real-time reverse transcription PCR (RT-PCR) strategy.
£217.00 - £1260.00
Specifications
Description | Works on all AB platforms, but is optimized for the 7500. |
---|---|
Detection Method | Primer-probe |
Format | Tube |
GC-Rich PCR Performance | High |
PCR Method | 1-step RT-qPCR |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
30057764
|
Applied Biosystems™
4387424M |
500 Reactions |
£678.00
500 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30057784
|
Applied Biosystems™
4387391M |
1000 Reactions |
£1260.00
1000 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30057797
|
Applied Biosystems™
AM1005M |
100 Reactions |
£217.00
100 reactions |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Recommended for RNA pathogen amplification
- Consistent amplification of RNA targets with high specificity and sensitivity
- Optimized to work with your target-specific primers and probes
- Contains ROX for quantitative fluorescent signal normalization
AgPath-ID™ One-Step RT-PCR Reagents are configured for fast and simple reaction setup. The reactions are assembled in a single tube, minimizing sample handling errors and expediting set-up time. Once assembled, results are available in approximately one hour. The 25X RT-PCR Enzyme Mix included in the kit contains highly efficient ArrayScript™ Reverse Transcriptase, a mutant MMLV RT that produces high cDNA yields, and AmpliTaq Gold™ polymerase, the preferred hot-start DNA polymerase for specific target amplification. The 2X RT-PCR Buffer has been optimized for efficient, robust reverse transcription and PCR includes the passive reference dye, ROX™ Dye, for quantitative fluorescent signal normalization.
Specifications
Works on all AB platforms, but is optimized for the 7500. | |
Tube | |
1-step RT-qPCR | |
Standard | |
ROX (Pre-mixed) | |
One-Step RT-PCR Reagent | |
25X |
Primer-probe | |
High | |
AmpliTaq Gold DNA Polymerase | |
7500 System | |
AgPath-ID™, Ambion™ | |
ArrayScript™ | |
25 X |
For Research Use Only. Not for use in diagnostic procedures.