missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AIM 100
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent Ack1 (TNK2) inhibitor (IC50 = 22 nM). Exhibits selectivity for Ack1 over ABL1, BTK, Lck and LYN; exhibits no inhibition of 25 other kinases. Suppresses phosphorylation of Tyr267 of the androgen receptor in prostate cancer cells. Also suppresses growth of radioresistant castration-resistant prostate cancer in xenograft tumors in mice.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 371.43 |
| Molecular Weight (g/mol) | 371.44 |
| Chemical Name or Material | AIM 100 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction