missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AIM V™ Medium, liquid (research grade)

AIM V™ Medium, liquid (research grade)
£82.50 - £128.00
Specifications
Cell Type | Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes |
---|---|
Form | Liquid |
Product Type | Serum Free Medium (SFM) |
With Additives | Glutamine, Phenol Red, HSA |
Product Line | AIM V™ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11590416
![]() |
Gibco™
12055091 |
500 mL |
£82.50
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10297582
![]() |
Gibco™
12055083 |
1000 mL |
£128.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Contains L-glutamine, streptomycin sulfate at 50μg/mL, and gentamicin sulfate at 10μg/mL.Specifications
Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes | |
Serum Free Medium (SFM) | |
AIM V™ | |
Streptomycin Sulfate, Gentamicin | |
Store in refrigerator (2–8°C) and protect from light. |
Liquid | |
Glutamine, Phenol Red, HSA | |
Room Temperature | |
Research Grade | |
Mammalian Cell Culture |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Gibco™ AIM V™ Medium, liquid (research grade)