missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ AIM V™ Medium (research grade), AlbuMAX™ Supplement
GREENER_CHOICE

Product Code. 11584416
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11584416

Brand: Gibco™ 31035025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

AIM V™ Medium (research grade), AlbuMAX™ Supplement

Contains AlbuMAX™ supplement (bovine serum albumin) substituted for human serum albumin. Contains L-glutamine, streptomycin sulfate at 50μg/mL, and gentamicin sulfate at 10μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Monocytes, Dendritic Cells, T Cells, Hybridomas, PBMCs, Fibroblasts, Macrophages, Myeloma Cells, Lymphocytes
Form Liquid
Product Type AIM V™ Medium
Sterility Sterile-filtered
With Additives Glutamine, Phenol Red, AlbuMAX
Product Line AIM V
Quantity 500 mL
Shipping Condition Room Temperature
Included Antibiotics Streptomycin Sulfate, Gentamicin
Purity or Quality Grade Research Grade
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.