missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIP/ARA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18176-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.Specifications
AIP/ARA9 | |
Polyclonal | |
Unconjugated | |
AIP | |
AIP, ARA9, FKBP16, FKBP37, HBV X associated protein 2, Immunophilin homolog ARA9, XAP 2, XAP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11632, 282827, 9049 | |
-20°C | |
Liquid |
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O00170, O08915, Q5FWY5 | |
Aip | |
AIP/ARA9 Fusion Protein Ag12869 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction