Learn More
Tuttnauer Air Compressor
Product Code. 17849538 Shop All Tuttnauer Products
Click to view available options
Frequency:
50 Hz
60 Hz
Unit Size:
Each
Product Code. 17849538
Brand: Tuttnauer PUM0590013
Only null left
Add to basket
Add to basket
The Air Compressor introduces pressurized air into the chamber for rapid cooling of liquids for models with a fast cooling system. Not needed if air pressure source is available on site.
  • Introduces pressurized air into the chamber for rapid cooling of liquids for models with a fast cooling system.
  • Not needed if air pressure source is available on site.
Specifications
Product Type Air Compressor
Frequency 60 Hz