missing translation for 'onlineSavingsMsg'
Learn More

Kimberly-Clark Professional™ Melodie™ Fragrance Clear Refill

Melodie™ Fragrance Clear Refill eliminates unwanted smells and improves user experience of any environment with floral, spicy notes.

Brand:  Kimberly-Clark Professional™ 6135

89.11 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.



Product Code. 10155987

  • £147.00 / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Compatible with the discontinued range aircare dispensers
  • Promotes hygiene
  • Improves comfort and care
  • Helps to manage costs through controlled dispensing
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Air Freshener Refill
310 mL
310 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Kimberly-Clark Professional™ Melodie™ Fragrance Clear Refill > Volume: 310 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.