Learn More
Cole-Parmer™ Air Lubricator
Dispenses oil mist at less than 1 cfm
Brand:  Cole-Parmer™ 07042-70
Product Code. 11729559
  • £102.24 / Each
Estimated Shipment: 19-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Air lubricator
  • Uses light spindle oil
  • Porous bronze wick
  • Measures 4 1/4 x 1 5/8 in. dia.
Specifications
Specifications
Air lubricator
1 to 20 scfm
1/4 in. NPT(F)
1/4 in. NPT(F)