missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Air-Powered Double-Diaphragm PP Pump

Safely handles abrasive fluids and slurries — ideal for high-viscosity and shear sensitive materials

Brand:  Cole-Parmer™ 74000-70

Product Code. 15223295

  • £736.00 / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Air-powered motor eliminates sparking concerns associate with electrical pumps
  • Bolted pump design allows easy access to pump head for maintenance and maintains seal after repeated servicing
  • Non-metallic pumps are ideally suited for highly corrosive fluids intermittent/on-demand, portable low abrasive fluids, and suspended solids
  • Encapsulated muffler provides excellent sound dampening
  • These pumps can move high-viscosity fluids such as paint, sludge, and sewage. However, the flow rate and pressure depend on the application.
Specifications

Specifications

15.1 L/min.
17.78 cm
66°C
Polypropylene, Santoprene
19.84 cm
Air-Powered Double-Diaphragm PP Pump, Santoprene; 4 GPM
13.97 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers