Learn More
Asecos™ Air Purifier PURIFIAIR.488
Product Code. p-7221813 Shop All asecos Products
Click to view available options
Product Type:
Air Purifier
Replacement Filter
Unit Size:
Each
Product Code. 17200197
Brand: Asecos™ 39311
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Air Purifier PURIFIAIR.488 reduces indoor pollutant levels with a multi-component filter system.
  • Asecos PURIFIAIR.488 can be used to reduce pollutant levels in indoor atmospheres
    • Removes mould spores, pollen, and allergens
    • Eliminates germs and bacteria and neutralises them
    • Continuously filters out fine dust particles
    • Removes unpleasant odours and harmful gases from indoor atmospheres
    • Built-in PM2.5 sensor and colour display allow users to quickly identify and easily read off the particle concentration in the indoor atmosphere in four levels from green (low) to red (high)
  • 1 device handles a volume flow rate of up to 488 m3/h and is designed for approx. 80 m3 room volume (corresponding to a surface area of 32 m2; at a room height of 2.5 m)
    • A larger number of devices should be used in larger rooms
    • Fresh air must be supplied to the rooms via natural or technical ventilation
    • Air purifiers should not be considered a substitute for a fresh air supply
    • Efficient results over time only if the filter is replaced at regularly according to the electronic display
    • Large filter units ensure a long service life
    • Low energy requirements result in low usage costs
    • Integrated humidification for a healthy and comfortable indoor climate
Specifications
Filter Type Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter
Airflow Rate 488 3/h
Weight (Metric) 11.3 Kg
Height (Metric) 669 mm
Depth (Metric) 400 mm
Width (Metric) 400 mm
Product Type Air Purifier
Voltage 220/240 V
Frequency 50/60 Hz
Certifications/Compliance CE Conformity