missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ HPLC Air Valve V2.0

Air valves are part of the SCAT™ Safety Cap system used to protect solvents in supply bottles from external contamination. They provide safe pressure equalization with clean air for your solvent inlet.

Brand:  SCAT™ 397010

190.41 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.



Additional Details : Weight : 0.03600kg

 View more versions of this product

Product Code. 16433819

  • £257.00 / Pack of 10
In Stock
to see stock.
Add to basket

Description

Description

An exchange indicator shows the elapsed service life, allowing for prompt replacement of the air valve after 6 months of use. Purified ventilation enables precise and reproducible analytical results in the HPLC/UHPLC process.

  • Blocks solvent vapors
  • Protects health and the environment
  • Exchange indicator displays next replacement time
  • Easy to change without tools
  • Fits any SCAT™ Safety Cap
Specifications

Specifications

10 Pc.
V2.0
10 Valves
0.25 in.
UNF 0.25 in., 28 G
Red
Polyurethane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers