Learn More
3M™ Aircare™ Filtration Unit ACU-03
3M™ Aircare™ Filtration Unit ACU-03 provides high quality air from an external source of compressed air by removing water and oil droplets, solid particles, and odors.
Brand:  3M™ 7100035217
Product Code. 15730058
  • £875.00 / Each
Estimated Shipment: 06-08-2025
to see stock.
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Supplies the highest quality air to the Supplied Air Respirator System and compressed air tools by removing water and oil aerosols, particles and odors from the facility supplied compressed air
  • Wall mounted unit
  • Can be used for up to three users or two users plus air tool
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CE Approved
ACU-03
Wall-mounted Aircare Filtration Unit
Gray
0.333 x 0.233 x 0.18 m
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Aircare™ Filtration Unit ACU-03 > Dimensions (L x W x H): 0.333 x 0.233 x 0.18 m