missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ AirClean™ AC600 Series ISO5/Class 100 Vertical Laminar Flow Workstation with UVTect™ Microprocessor Controller
GREENER_CHOICE

The AirClean™ AC600 Series ISO5/Class 100 Vertical Laminar Flow Workstation with UVTect™ Microprocessor Controller is ideal for non-hazardous applications that require a clean work area. These compact, lightweight workstations provide positive-pressure, ISO 5 air.

Brand:  Syngene™ SYN-AC648LFC220-E

6330.00 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.


Product Code. 17765904

  • £7610.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

AirClean™ Systems AC600 Series Vertical Laminar Flow Workstations are ideal for non-hazardous applications that require a clean work area. These compact, lightweight workstations provide positive-pressure, ISO 5 air throughout the chamber and sash opening, thus keeping airborne particulate away from sensitive applications. Given the small footprint and ease of installation (simply plug in the power cord), the AC600 Series Vertical Laminar Flow Workstations are an excellent choice for mobile ‘clean zones’ within your facility.

Standard Features
  • No ductwork required
  • No installation required - plugs directly into a standard 110V or 220V electrical outlet
  • Clear polycarbonate shell for 360° visibility
  • Available in standard height or tall version
  • Polypropylene construction for chemical resistance. The result: NO RUST
  • UVTect™ Microprocessor Controller
  • ISO 5 vertical laminar flow air
  • Lab event timer
  • Integral fluorescent lighting
Unique Safety Features
  • Filter safety lockout to ensure correct filter replacement
  • Shipped fully assembled
  • Tested to UL 61010-1, CSA C22.2 No. 61010-1, and EN 61326-1
  • Filter and electronics are located above the workspace, eliminating the chance of chemical spill
  • ISO 5 tested
  • Real time monitoring of installed filtration bed
Specifications

Specifications

Polypropylene
32 in.
81 cm
UVTect™ Microprocessor Controller
1220 mm
UL 61010-1, CSA C22.2 No. 61010-1, and EN 61326-1
18 in.
46 cm
48 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138

    Expires2025-04-30

  • Multiple Manufacturers
    Up to 60% off
    Promo Code24119

    Expires2025-04-30

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Syngene™ AirClean™ AC600 Series ISO5/Class 100 Vertical Laminar Flow Workstation with UVTect™ Microprocessor Controller > Vertical Laminar Flow; Width: 48 in.; Includes: UVTect™ Microprocessor Controller

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.