missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ AirClean™ Blended Bonded Carbon Filter for Acids and Formaldehyde

The AirClean™ Blended Bonded Carbon Filter for Acids and Formaldehyde is used to absorb fumes and odors.

Brand:  Syngene™ SYN-ACF4F4

Additional Details : Weight : 3.62870kg

 View more versions of this product

Product Code. 17795964

  • £1200.00 / Each
Estimated Shipment: 22-08-2024
to see stock.

Description

Description

AirClean™ Systems manufactures gas phase bonded carbon and multi-layered filters that are available with standard coconut shell carbon or chemically impregnated carbon, depending on your application. AirClean™ Systems gas-phase, coconut shell carbon filters utilize our unique bonded carbon filtration media. This minimizes the release of potentially hazardous carbon dust found in traditional granular carbon filters. Our proprietary bonding process holds carbon in a predictable matrix, preventing carbon shift that can lead to 'dead spots', common in traditional granular filters.

Features

  • Bonded carbon — no filter dusting
  • Even distribution of impregnates, if applicable
  • Multi-layering options for larger application compatibility
Specifications

Specifications

Carbon
3 in.
1 Ea.
30.5 cm
Acids, Formaldehyde
12 in.
24 in.
8 cm
61 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers