missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Airclean™ Process Protection Microscope Hood
GREENER_CHOICE

Product Code. p-7193308 Shop All Syngene Products
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16532972

Brand: Syngene™ SYN32PPUVCE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Process Protection with UVTect™ Microprocessor and Microscope Sash

  • AirClean™ Systems AC600 Series Vertical Laminar Flow Workstation
  • Laminar air flow is described as the smooth, fluid, parallel flow of air in a consistent direction
  • Ideal for applications that are not hazardous to the operator but require a clean workspace
  • A compact, light-weight design allows for multiple clean zones within a laboratory
  • Two-stage filtration provides clean, Class 100 laminar flow air for process protection
  • Room air first passes through an electrostatically charged pre-filter followed by a HEPA filter resulting in ISO 5 clean air
  • Independently certified to meet or exceed applicable clean bench standards

Specifications

Product Type Laminar Flow
Depth (Metric) 61 cm
Height (Metric) 76.2 cm
Width (English) 32 in.
Width (Metric) 81.28 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.