missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Black
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
£26.25 - £59.60
Specifications
Color | Black |
---|---|
Material | Microcellular Vinyl |
Type | Industrial and Commercial Matting |
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Price | Quantity & Availability | ||||
17722068
|
Notrax
410S0323BL |
91 cm | 60 cm |
£26.25
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
17752068
|
Notrax
410S0335BL |
150 cm | 91 cm |
£59.60
Each |
Estimated Shipment: 07-01-2025
Log in to see stock. |
||||
17732058
|
Notrax
410C0324BL |
1830 cm | 60 cm | N/A | |||||
17762058
|
Notrax
410C0336BL |
1830 cm | 91 cm | N/A | |||||
17792058
|
Notrax
410C0348BL |
1830 cm | 122 cm | N/A | |||||
Description
- Ribbed pattern enhances traction
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
Black | |
Microcellular Vinyl | |
Industrial and Commercial Matting |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title