missing translation for 'onlineSavingsMsg'
Learn More

Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Black/Yellow

Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.

Brand:  Notrax 410C0348BY

Additional Details : Weight : 2.80000kg

 View more versions of this product

Product Code. 17702068

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Ribbed pattern enhances traction
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Specifications

Specifications

60 ft.
Industrial and Commercial Matting
122 cm
Black/Yellow
Microcellular Vinyl
1830 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers