missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Notrax Airug™ Construction of Microcellular Vinyl with Ribbed Pattern, Gray
Notrax Airug™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles.
Brand: Notrax 410C0336GY
Additional Details : Weight : 2.10000kg
Description
- Ribbed pattern enhances traction
- Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
- All four sides are bevelled and sealed to minimize tripping hazards
Specifications
60 ft. | |
Industrial and Commercial Matting | |
91 cm |
Gray | |
Microcellular Vinyl | |
1830 cm |