missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AK 7, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective SIRT2 inhibitor (IC50 = 15.5 μM); displays no effect on SIRT1 or SIRT3. Decreases neuronal cholesterol levels; improves motor function and ameliorates brain atrophy in a mouse model of Huntington's disease. Brain penetrant.
Specifications
Quantity | 50 mg |
Formula Weight | 437.35 |
Percent Purity | >98% |
Chemical Name or Material | AK 7 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'