missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies AKT Inhibitor VIII
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Description
- AKT Inhibitor VIII is a cell-permeable, allosteric inhibitor of all three forms of the kinase AKT (AKT1, AKT2, and AKT3) with IC50 values of 58, 210, and 2,200 nM, respectively (Lindsley et al.; Calleja et al.) It displays good selectivity against a panel of 70 other kinases with micromolar inhibition against some kinases, such as calcium/calmodulin-dependent protein kinase 1 and smooth muscle myosin light-chain kinase (Logie et al.)
- Sensitizes prostate tumor and cervical carcinoma cells to apoptotic stimuli (DeFeo-Jones et al.)
- Blocks mitosis and inhibits migration of HeLa cells (Jo et al.)
- Reduces insulin-dependent gene repression in liver cells leading to reduced insulin sensitivity (Logie et al.)
Specifications
Specifications
| CAS | 612847-09-3 |
| Synonym | AKTi-1/2; AKT 1/2 inhibitor |
| Quantity | 1 mg |
| Molecular Formula | C34H29N7O |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction