missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 0.6mm, I.D.: 0.3mm, Thermo Scientific Chemicals

£17.30 - £36.73

Specifications

Chemical Name or Material Al-23 Insulating Tube
Name Note Both ends open (Thin Wall)
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11337478
View Documents
Thermo Scientific Alfa Aesar
031476.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11357478
View Documents
Thermo Scientific Alfa Aesar
031476.DR
200 mm
£29.09
200mm
Please to purchase this item. Need a web account? Register with us today!
11367478
View Documents
Thermo Scientific Alfa Aesar
031476.DT
250 mm
£36.73
250mm
Please to purchase this item. Need a web account? Register with us today!
11347478
View Documents
Thermo Scientific Alfa Aesar
031476.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Al-23 Insulating Tube
Both ends open (Thin Wall)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only