missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Al-23 Solid Round Rod, Dia.: 0.6mm, Thermo Scientific™
£12.40 - £26.72
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323038
|
Thermo Scientific Alfa Aesar
032921.DM |
100 mm |
£12.40
100mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333038
|
Thermo Scientific Alfa Aesar
032921.DR |
200 mm | |||||||
11343038
|
Thermo Scientific Alfa Aesar
032921.DW |
300 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Al-23 Solid Round Rod |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures