missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, Both Ends Open, O.D.: 6mm, I.D.: 3mm, Thermo Scientific™

£23.00 - £99.00

Specifications

Chemical Name or Material Al-23 Tube
Name Note Both ends open
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11393927
View Documents
Thermo Scientific Alfa Aesar
031556.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11383927
View Documents
Thermo Scientific Alfa Aesar
031556.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11323937
View Documents
Thermo Scientific Alfa Aesar
031556.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11363927
View Documents
Thermo Scientific Alfa Aesar
031556.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11313937
View Documents
Thermo Scientific Alfa Aesar
031556.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373927
View Documents
Thermo Scientific Alfa Aesar
031556.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Al-23 Tube
Both ends open
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications