missing translation for 'onlineSavingsMsg'
Learn More

Al-23 Tube, One End Closed, O.D.: 30mm, I.D.: 25mm, Thermo Scientific Chemicals

POA - £300.50

Specifications

Chemical Name or Material Al-23 Tube
Name Note One end closed
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352808
View Documents
Thermo Scientific Alfa Aesar
030080.DM
100 mm
£257.28
100mm
Please to purchase this item. Need a web account? Register with us today!
11322808
View Documents
Thermo Scientific Alfa Aesar
030080.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11342808
View Documents
Thermo Scientific Alfa Aesar
030080.DW
300 mm
£212.06
300mm
Please to purchase this item. Need a web account? Register with us today!
11332808
View Documents
Thermo Scientific Alfa Aesar
030080.EH
600 mm
£300.50
600mm
Please to purchase this item. Need a web account? Register with us today!
11302808
View Documents
Thermo Scientific Alfa Aesar
030080.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Al-23 Tube
One end closed
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only