Learn More
Thermo Scientific™ alamarBlue™ Dye
Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
Brand:  Thermo Scientific™ Y00-100
360.55 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15559604
  • £545.00 / 100mL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alamarBlue indicator dye
Indicator Dye
100 mL