missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals β-Alanine, 99% Product Code.: 10245880

Thermo Scientific Chemicals β-Alanine, 99%

Product Code. 10245880
500 g, Plastic Bottle
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic Drum
This item is not returnable. View return policy

Product Code. 10245880

Brand: Thermo Scientific Chemicals 163795000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-95-9
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00008200
InChI Key UCMIRNVEIXFBKS-UHFFFAOYSA-N
Synonym beta-alanine, 3-aminopropionic acid, abufene, beta alanine, h-beta-ala-oh, beta-aminopropionic acid, 2-carboxyethylamine, .beta.-alanine, alanine, beta, propanoic acid, 3-aminoShow MoreShow Less
PubChem CID 239
ChEBI CHEBI:16958
IUPAC Name 3-aminopropanoic acid
SMILES NCCC(O)=O

Specifications

Melting Point 197.0°C to 202.0°C
Ammonium 1000ppm max.
CAS Min % 98.5
CAS Max % 100.0
pH 6.5 to 7.5 (10% w/v aq. soln.)
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Other Amino Acids (TLC) Passes Test (not detectable on 10μgspotting)
Packaging Plastic Bottle
Linear Formula H2NCH2CH2CO2H
Fieser 01,16; 05,6; 06,10; 17,3
Sulfate 480ppm max.
Sulfated Ash 0.2% max.
Quantity 500 g
Formula Weight 89.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material β-Alanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458278 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number163795000,163790025,163790100
Lot NumberA0467286 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number163795000,163790025,163790100
Lot NumberA0462776 Certificate TypeCertificate of Analysis Date16/05/2024 Catalog Number163795000,163790025,163790100
Lot NumberA0460095 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number163795000,163790025,163790100
Lot NumberA0450058 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number163795000,163790025,163790100
Product Identifier
  • ß-Alanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals β-Alanine, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.