Learn More
Emma Safety Footwear Alaska XXD Safety Shoes
Offers exceptional comfort while the steel toe cap protects from falling objects. Easy twist sole provides a 'sticky grip'.
Brand:  Emma Safety Footwear MM30603641
 View more versions of this product
Product Code. 15948341
  • £144.16 / 1 pair
Estimated Shipment: 15-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Specifications
EN ISO 20345:2011, EN 12568:2010
Black
Hydro-Tec™ Sanitized Silver
41
Laces
Alaska XXD
Oiled Nubuck
Steel