missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ AlbuMAX™ II Lipid-Rich BSA
AlbuMAX™ II Lipid-Rich BSA
£107.00 - £3010.00
Specifications
Content And Storage | Store in refrigerator (2–8°C). |
---|---|
Product Type | BSA |
Product Line | AlbuMAX™, Gibco™ |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11550376
|
Gibco™
11021029 |
25 g |
£107.00
25g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11560376
|
Gibco™
11021037 |
100 g |
£393.00
100g |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10624053
|
Gibco™
11021045 |
1 kg |
£3010.00
1kg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Lipid-rich bovine serum albumin for cell culture. It is chromatographically purified and has an IgG content ≤1.0%.Origin:
New Zealand
Specifications
Store in refrigerator (2–8°C). | |
BSA | |
AlbuMAX™, Gibco™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.