missing translation for 'onlineSavingsMsg'
Learn More

Albumin Mouse anti-Human, Porcine, Rat, Clone: 4A1C11, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66051-1-IG-150UL

Product Code. 16877575

This item is not returnable. View return policy

Description

Description

Albumin is a soluble, monomeric protein which comprises about one-half of the blood serum protein. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular unglycosylated serum protein of molecular weight 65, 000. Albumin is synthesized in the liver as preproalbumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough endoplasmic reticulum. The product, proalbumin, is in turn cleaved in the Golgi vesicles to produce the secreted albumin.
Specifications

Specifications

Albumin
Monoclonal
0.71 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02768, P02770, P08835
ALB
Albumin Fusion Protein Ag9885
150 μL
Primary
Human, Porcine, Rat
Antibody
IgG1
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
4A1C11
Unconjugated
ALB
Alb, albumin, Serum albumin
Mouse
Protein A
RUO
213, 24186, 396960
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers