Learn More
Thermo Scientific Chemicals Alcian Blue 8GX
Description
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alcian Blue 8GX is widely utilized as a heteroglycan stain for acidic, neutral, sulfated and phosphate polysaccharides and glycosaminoglycans in tissues. It finds application in both light and electron microscopy. It serves as an adhesive to stick glycol methacrylate sections to glass slides. Due to its large aromatic surface, it is used as a coating agent and modifies the property on the surfaces. It acts as a gelling agent for lubricating fluids and is also used in the modification of carbon paste electrode which promotes redox behavior and stability of electrode.
Solubility
Soluble in water, ethanol, ethylene glycol and 2-ethoxyethanol. Insoluble in xylene.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 148°C |
Merck Index | 14,218 |
Solubility Information | Soluble in water,ethanol,ethylene glycol and 2-ethoxyethanol; Insoluble in xylene. |
Formula Weight | 1298.88 |
Color | Blue |
Quantity | 10 g |
Physical Form | Powder |
Chemical Name or Material | Alcian Blue 8GX |
RUO – Research Use Only