missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Alcian Blue 8GX, certified

Product Code. 10619263
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33864-99-2
Molecular Formula C56H68Cl4CuN16S4
Molecular Weight (g/mol) 1298.86
MDL Number MFCD00010720
InChI Key XRWQTDIELZDCPS-UHFFFAOYSA-J
Synonym Ingrain Blue 1
PubChem CID 17749099
IUPAC Name (9Z,18Z,26Z,35Z)-4,14,22,32-tetrakis({[(dimethylamino)(dimethyliminiumyl)methyl]sulfanyl}methyl)-9,18,27,36,37,39,40,41-octaaza-38-cupradecacyclo[17.17.3.1¹⁰,¹⁷.1²⁸,³⁵.0²,⁷.0⁸,³⁷.0¹¹,¹⁶.0²⁰,²⁵.0²⁶,³⁹.0²⁹,³⁴]hentetraconta-1,3,5,7,9,11(16),12,14,17(41),18,20,22,24,26,28(40),29(34),30,32,35-nonadecaene-38,38-bis(ylium) tetrachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].CN(C)C(SCC1=CC2=C(C=C1)\C1=N\C3=C4C=CC(CSC(N(C)C)=[N+](C)C)=CC4=C4\N=C5/N=C(/N=C6\N([Cu++]N34)\C(=N/C2=N1)C1=CC(CSC(N(C)C)=[N+](C)C)=CC=C61)C1=C5C=C(CSC(N(C)C)=[N+](C)C)C=C1)=[N+](C)C

Specifications

Melting Point 148.0°C
Dye Content 50% min.
Merck Index 15, 212
Solubility Information Solubility in water: moderately soluble. Other solubilities: soluble in ethanol, cellosolve, ethylene glycol, practically insoluble in xylene
Infrared Spectrum Authentic
Formula Weight 1298.88
Packaging Glass bottle
Color Purple
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Alcian Blue 8GX
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'