missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Alcian Blue/PAS Special Stain Kit
Demonstrate the presence of both neutral and acidic mucosubstances with the Epredia™ Alcian Blue/PAS Special Stain Kit
Brand: Epredia™ 88016
86.94 GBP valid until 2024-10-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections
- Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances
Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent
Specifications
Periodic Acid Schiff |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- periodic acid solutiuon (0.5-1%)
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362-Take off contaminated clothing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : periodic acid