missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Alcian Blue/PAS Special Stain Kit

Product Code. p-4530172 Shop All Epredia Products
Click to view available options
Type:
Alcian Blue
Alcian Blue/PAS Special Stain Kit
Periodic Acid Schiff
Unit Size:
500mL
Pack of 3
This item is not returnable. View return policy

Product Code. 12666387

Brand: Epredia™ 87023

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Demonstrate the presence of both neutral and acidic mucosubstances with the Epredia™ Alcian Blue/PAS Special Stain Kit

  • Can be used alone to stain acid mucins and other carboxylated and sulfated acid mucosubstances blue on tissue sections
  • Periodic Acid Schiff (PAS) reaction is used to stain glycogen, basement membranes and neutral mucosubstances

Includes: Alcian Blue pH 2.5 stain solution, periodic acid solution (0.5%) and Schiff reagent

TRUSTED_SUSTAINABILITY

Specifications

Type Alcian Blue/PAS Special Stain Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Alcian Blue/PAS Kit 87023
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362-Take off contaminated clothing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : periodic acid, 2-methyl-3-isothiazolone,acetic acid, 5-chloro-2-methyl-3-isothiazolone, copper(4+)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.