missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alcohol, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£805.00
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | methanol |
SMILES | CO |
Description
- For Histological Use; NOTE: Intended for nonfuel end purposes only
Chemical Identifiers
64-17-5 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
702 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:16236 | |
CO |
Specifications
64-17-5 | |
1L = 0.79kg | |
MFCD00004595 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
702 | |
Meets ACS reagent chemical requirements |
13.00°C (55.4°F) | |
CH4O | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:16236 | |
Alcohol, Anhydrous |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H371-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.